EasySep™ Mouse NK Cell Isolation Kits
Lieferant: STEMCELL Technologies
19855 19855RF
STMC19855EA 1450 EUR
STMC19855 STMC19855RF
EasySep™ Mouse NK Cell Isolation Kits
Zelltrennmedien
Immunomagnetic isolation of untouched mouse NK cells
  • Fast and easy-to-use
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet. Desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays.
This product replaces the EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755) for even faster cell isolations.
Bitte beachten Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').