Human Recombinant EPO (CHO-expressed)
Lieferant: STEMCELL Technologies
78007 78007.1 78007.2
STMC78007EA 292 EUR
STMC78007 STMC78007.1 STMC78007.2
Human Recombinant EPO (CHO-expressed)
Proteine und Peptide
The principal physiological regulator of erythropoiesis, Erythropoietin (EPO) is a glycoprotein growth factor that is produced primarily in the kidney in response to hypoxia or anemia. EPO promotes erythropoiesis by binding to a homodimeric cell surface receptor that activates JAK2/STAT5, PI3K/AKT, and MAPK pathways, and stimulates the proliferation and differentiation of erythroid progenitor cells (Jelkmann. Transfus Med Hemother, 2013; Kuhrt and Wojchowski. Blood, 2015).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').