Human Recombinant Shh (C24II)
Lieferant: STEMCELL Technologies
78065 78065.1 78065.2
STMC78065EA 650 EUR
STMC78065 STMC78065.1 STMC78065.2
Human Recombinant Shh (C24II)
Proteine und Peptide
Sonic hedgehog (Shh) is a member of the Hedgehog family of secreted signaling proteins that has a prominent role in patterning during early mammalian development (Ho and Scott). It is expressed during early embryogenesis in a variety of tissues (Finco et al.). During patterning of the central nervous system, Shh directly acts on cells to specify neural cell fates, regulates proliferation and survival of oligodendrocytes precursors and neural crest cells and stimulates proliferation, differentiation and axon growth (Ho and Scott; Martí). Shh also regulates T cell differentiation and activation (Crompton et al.). Recombinant human Shh (C24II; rhShh) is a fully biologically active molecule.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').