Human Recombinant IL-6R alpha
Lieferant: STEMCELL Technologies
78083.1
STMC78083.1EA 881 EUR
STMC78083.1
Human Recombinant IL-6R alpha
Proteine und Peptide
Interleukin 6 receptor (IL-6R) alpha is a type I transmembrane glycoprotein that forms a complex with type I transmembrane signal transducer protein gp130 (CD130) and mediates the biological activities of IL-6. IL-6 binds to the membrane-bound non-signaling IL-6R alpha (mIL-6R), and the complex binds to two molecules of gp130 and leads to ‘classical’ IL-6-signal transduction, which includes activation of JAK/STAT, ERK, and PI3K signal transduction pathways (Scheller et al.). In contrast, a soluble form of IL-6R alpha (sIL-6R), which comprises the extracellular portion of the receptor, binds to the secreted IL-6 to form a complex that promotes bioavailability of IL-6. The complex of IL-6 and sIL-6R can bind to gp130 on cells that do not express the IL-6R and are unresponsive to IL-6. This process is known as trans-signaling (Hunter and Jones; Rose-John S). sIL-6R regulates both local and systemic IL-6-mediated events. Elevated levels of sIL-6R have been documented in several disease conditions such as rheumatoid arthritis, myeloma, and Crohn’s disease (Jones et al.; Mihara et al.).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').