Human Recombinant IL-9
Lieferant: STEMCELL Technologies
Interleukin 9 (IL-9) has pleiotropic functions in the immune system and signals through its specific IL-9 receptor (IL-9R) (Renauld et al.). IL-9/IL-9R signaling pathway mainly targets the downstream activation of JAK/STAT (janus kinase/signal transducer and activator of transcription) and subsequent phosphorylation cascades initiated by multiple kinases including IRS–PI3K–PKB (insulin receptor substrate, phosphatidyl-inositol 3-kinases, protein kinase-B) and ERK (Knoops and Renauld; Fontaine et al.). IL-9 has been shown to have a role in Th1/Th17-mediated inflammation and in regulatory T cell responses (Singh et al.; Goswami and Kaplan). IL-9/IL-9R signaling pathway represents a novel endogenous anti-apoptotic mechanism for cortical neurons (Fontaine et al.). IL-9 secreting T cells, termed Th9 cells, contribute to both effective immunity and immunopathological disease, and have been shown to have a role in the treatment of allergic and autoimmune disease (Kaplan et al.).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').