Human Recombinant MCP-1 (CCL2)
Lieferant: STEMCELL Technologies
78087 78087.1
STMC78087EA 120 EUR
STMC78087 STMC78087.1
Human Recombinant MCP-1 (CCL2)
Proteine und Peptide
Monocyte chemotactic protein-1 (MCP-1), also known as CCL2, is a member of the CC family of chemokines. The protein is primarily induced by platelet-derived growth factor (PDGF) gene (Cochran et al.). The biological effects of MCP-1 are mediated via the specific G-protein-coupled receptor CCR2 which in turn activates signal transduction pathways leading to monocyte transmigration (Sozzani et al.). Migration of monocytes from the bloodstream across the vascular endothelium is required for routine immunological surveillance of tissues, as well as other immunomodulatory effects. MCP-1 is produced by a variety of cell types, including fibroblasts and endothelial, epithelial, smooth muscle, mesangial, astrocytic, monocytic, and microglial cells, which are important for antiviral responses in the peripheral circulations and in tissues (Cushing et al.; Deshmane et al.). MCP-1 plays a role in physiological processes such as neurogenesis, neuroprotection, and neurotransmission and has important implications in neurological disorders such as multiple sclerosis and Alzheimer’s disease, in which it is produced during neuroinflammation at the sites of lesions (Conductier et al.).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').