Human Recombinant FGF-4
Lieferant: STEMCELL Technologies
78103.1 78103.2
STMC78103.1EA 282 EUR
STMC78103.1 STMC78103.2
Human Recombinant FGF-4
Proteine und Peptide
Fibroblast growth factor 4 (FGF-4) is a member of the FGF superfamily (Beenken and Mohammadi). FGF-4 is expressed in pluripotent stem cells and is implicated in various stages of development and morphogenesis in a variety of organisms (Kosaka et al. 2009). FGF-4 has been shown to have an ability to promote neural stem cell proliferation and neuronal differentiation in the postnatal brain (Kosaka et al. 2006). FGF-4 has also been shown to increase the proliferation rate of mesenchymal stem cells (Farré et al.). FGF-4 supports the maintenance and self-renewal properties of human embryonic stem cells and also promotes the proliferation of these cells (Mayshar et al.). In the mouse, the Fgf4 gene also supports proliferation of the inner cell mass (ICM) and postimplantation embryos (Feldman et al.).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').