Human/Mouse Recombinant TGF-beta 3, ACF
Lieferant: STEMCELL Technologies
78156 78156.1
STMC78156EA 326 EUR
STMC78156 STMC78156.1
Human/Mouse Recombinant TGF-beta 3, ACF
Proteine und Peptide
Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes. TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.). TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.). TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.). This product is animal component-free.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Bestellen
TECHNISCHE DATEN
Protein/Peptid-Name TGF-beta 3, ACF
Environmentally Preferable
Protein/Peptid-Typ Rekombinant
Spezies Human,Mouse
Ursprung
Purity ≥95%