Mouse Recombinant IL-19
Lieferant: STEMCELL Technologies
78207 78207.1
STMC78207EA 274 EUR
STMC78207 STMC78207.1
Mouse Recombinant IL-19
Proteine und Peptide
Interleukin 19 (IL-19) is a member of the IL-10 cytokine family and is produced by keratinocytes, B cells, and monocytes (Romer et al.; Wolk et al.). Expression of IL-19 can be induced by granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS; Gallagher et al.). IL-19 is considered to be a proinflammatory cytokine, as it upregulates IL-6 and tumor necrosis factor alpha (TNF-α; Liao et al. 2002). IL-19 binds the IL-20 receptor complex (IL-20R) which comprises IL-20R alpha and IL-20R beta to activate the STAT3 pathway (Dumoutier et al.). IL-19 also induces T-helper cell differentiation towards a Th2 response, resulting in the production of IL-10 and additional IL-19 (Liao et al. 2002; Liao et al. 2004). IL-19 has been implicated in aging, vascular disease, Type I diabetes, and rheumatoid arthritis.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').