Human Recombinant RANKL
Lieferant: STEMCELL Technologies
78214 78214.1 78214.3
STMC78214EA 291 EUR
STMC78214 STMC78214.1 STMC78214.3
Human Recombinant RANKL
Proteine und Peptide
Receptor activator of nuclear factor kappa-B ligand (RANKL) is a member of the tumor necrosis factor (TNF) superfamily (Anderson et al.). Cytokines in the TNF superfamily are involved in a variety of long-term cellular activities such as differentiation, proliferation, and cell death (MacEwan). RANKL is a type II homotrimeric transmembrane protein expressed in both a membrane-bound and secreted form (Ikeda et al.). RANKL binds to the receptor activator of nuclear factor kappa-B (RANK). Upon binding to its receptor, RANKL activates the AKT signaling pathway (Moon et al.). Osteoprotegerin (OPG) may also bind RANKL, and this binding competes with RANKL-RANK binding (Lacey et al.). RANKL is involved in osteoclastogenesis (Lacey et al.; Yasuda et al.) and T cell activation (Wong et al.).
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').