Wasser nukleasefrei

Lieferant: STEMCELL Technologies

Synonyme: Wasserstoffoxid, Hydrogenoxid

79001 79002 79002.1
STMC79001EA 90.9 EUR
STMC79001 STMC79002 STMC79002.1
Wasser nukleasefrei
Wasser

DNase- and RNase-free water for molecular biology applications.


Nuclease-Free Water is used in various molecular biology applications requiring nuclease-free conditions, such as in processing DNA or RNA (i.e. PCR, cDNA synthesis, or qPCR). This product is sterile-filtered (0.2 µm), free of RNase and DNase activity, endotoxin-free, and not DEPC-treated.


Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR