MethoCult™ GF H84544 Medium
Lieferant: STEMCELL Technologies
Methylcellulose-based medium with recombinant cytokines (without EPO). 24×3 ml vials.
MethoCult™ GF H84534 is a methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples. H84534 has been formulated to support the optimal growth of granulocyte and macrophage progenitor cells (CFU-GM, CFU-G and CFU-M). This formulation is compatible with STEMvision™ software for automated colony counting.
Please Note: In light of the new EU Regulation 2017/746 (IVDR) of the European Parliament and of the Council on in vitro diagnostic medical devices, which is expected to become effective in 2022, STEMCELL recently deregistered and removed the CE IVD claims associated with these products, and they are now labeled “For Research Use Only”. Please note there have been no changes to the form, function and quality assurances associated with these products.
Bitte beachten Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').