Anti-CSNK2A1 Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
KAP-ST010E
STSSKAP-ST010EEA 0 EUR
STSSKAP-ST010E
Anti-CSNK2A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CSNK2A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR