Anti-NF-kappa B P50 Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
KAP-TF112C
STSSKAP-TF112CEA 0 EUR
STSSKAP-TF112C
Anti-NF-kappa B P50 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NF-kappa B P50
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR