Anti-BVR Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
OSA-400E
STSSOSA-400EEA 0 EUR
STSSOSA-400E
Anti-BVR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BVR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR