Anti-Glutathione Nem Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SPA-540C
STSSSPA-540CEA 0 EUR
STSSSPA-540C
Anti-Glutathione Nem Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: Glutathione Nem
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammals
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR