Anti-CRP55 Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SPA-600D
STSSSPA-600DEA 0 EUR
STSSSPA-600D
Anti-CRP55 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CRP55
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR