Anti-HSP25 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: G3.1]

Lieferant: Enzo Life Sciences (Stressgen)
SPA-800FID
STSSSPA-800FIDEA 0 EUR
STSSSPA-800FID
Anti-HSP25 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: G3.1]
Antikörper
Type: Primary
Antigen: HSP25
Clonality: Monoclonal
Clone: G3.1
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR