Anti-HSP70 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]

Lieferant: Enzo Life Sciences (Stressgen)
SPA-810FID
STSSSPA-810FIDEA 0 EUR
STSSSPA-810FID
Anti-HSP70 Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
Antikörper
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR