Anti-HSP70 Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SPA-811D
STSSSPA-811DEA 0 EUR
STSSSPA-811D
Anti-HSP70 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR