Anti-HSPA8 Rat Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SPA-815D
STSSSPA-815DEA 0 EUR
STSSSPA-815D
Anti-HSPA8 Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA8
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR