Anti-HSPA8 Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SPA-822D
STSSSPA-822DEA 0 EUR
STSSSPA-822D
Anti-HSPA8 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA8
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR