Anti-HSP90A Rat Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SPA-840D
STSSSPA-840DEA 0 EUR
STSSSPA-840D
Anti-HSP90A Rat Monoclonal Antibody
Antikörper
Type: Primary
Antigen: HSP90A
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR