Anti-DNAK Mouse Monoclonal Antibody () [clone: 8E2/2]

Lieferant: Enzo Life Sciences (Stressgen)
SPA-880D
STSSSPA-880DEA 0 EUR
STSSSPA-880D
Anti-DNAK Mouse Monoclonal Antibody () [clone: 8E2/2]
Antikörper
Type: Primary
Antigen: DNAK
Clonality: Monoclonal
Clone: 8E2/2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR