Anti-ESR Mouse Monoclonal Antibody [clone: h-151]

Lieferant: Enzo Life Sciences (Stressgen)
SRA-1000F
STSSSRA-1000FEA 0 EUR
STSSSRA-1000F
Anti-ESR Mouse Monoclonal Antibody [clone: h-151]
Antikörper
Type: Primary
Antigen: ESR
Clonality: Monoclonal
Clone: h-151
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR