Anti-FKBP59 Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SRA-1400D
STSSSRA-1400DEA 0 EUR
STSSSRA-1400D
Anti-FKBP59 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: FKBP59
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR