Anti-SYT1 Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
SYA-148D
STSSSYA-148DEA 0 EUR
STSSSYA-148D
Anti-SYT1 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SYT1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR