Anti-DLG4 Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
VAM-PS002C
STSSVAM-PS002CEA 0 EUR
STSSVAM-PS002C
Anti-DLG4 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR