Anti-SNAPA Mouse Monoclonal Antibody () [clone: 6B7-3]

Lieferant: Enzo Life Sciences (Stressgen)
VAM-SV027C
STSSVAM-SV027CEA 0 EUR
STSSVAM-SV027C
Anti-SNAPA Mouse Monoclonal Antibody () [clone: 6B7-3]
Antikörper
Type: Primary
Antigen: SNAPA
Clonality: Monoclonal
Clone: 6B7-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Sheep, Dog, Bovine, Hamster
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR