Anti-AMPH Mouse Monoclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
VAM-SV030C
STSSVAM-SV030CEA 0 EUR
STSSVAM-SV030C
Anti-AMPH Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: AMPH
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR