Anti-GANAB Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
VAP-PT034C
STSSVAP-PT034CEA 0 EUR
STSSVAP-PT034C
Anti-GANAB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GANAB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR