Anti-Membrin Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
VAP-PT049C
STSSVAP-PT049CEA 0 EUR
STSSVAP-PT049C
Anti-Membrin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Membrin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR