Anti-STX1A Rabbit Polyclonal Antibody

Lieferant: Enzo Life Sciences (Stressgen)
VAP-SV064E
STSSVAP-SV064EEA 0 EUR
STSSVAP-SV064E
Anti-STX1A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STX1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR