Mouse single tissue blot, single species
Lieferant: G Bioscienes
TB02 TB05 TB08 TB11 TB14 TB17 TB20 TB23 TB26
TB02EA 676 EUR
TB02 TB05 TB08 TB11 TB14 TB17 TB20 TB23 TB26
Mouse single tissue blot, single species
Elektrophorese-Reagenzien Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
  • Ready to screen
  • Single tissue mouse blots