Mouse single tissue blot, single species
Lieferant: G Bioscienes
TB02
TB05
TB08
TB11
TB14
TB17
TB20
TB23
TB26
TB02EA
676
EUR
TB02
TB05
TB08
TB11
TB14
TB17
TB20
TB23
TB26
Mouse single tissue blot, single species
Elektrophorese-Reagenzien
Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
- Ready to screen
- Single tissue mouse blots