Rat single tissue blot, single species
Lieferant: G Bioscienes
TB03 TB06 TB09 TB12 TB15 TB18 TB21 TB24 TB27
TB03EA 676 EUR
TB03 TB06 TB09 TB12 TB15 TB18 TB21 TB24 TB27
Rat single tissue blot, single species
Elektrophorese-Reagenzien Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
  • Ready to screen
  • Single tissue rat blots