Rat single tissue blot, single species
Lieferant: G Bioscienes
TB03
TB06
TB09
TB12
TB15
TB18
TB21
TB24
TB27
TB03EA
676
EUR
TB03
TB06
TB09
TB12
TB15
TB18
TB21
TB24
TB27
Rat single tissue blot, single species
Elektrophorese-Reagenzien
Western Blotting Reagents
This blot contains two protein lanes, a protein marker lane and a lane with one GenLysate™ sample. After the proteins are resolved, they are transferred to a PVDF membrane using Western Transfer Buffer. The blots are ready to be blocked and probed with the antibodies of choice.
- Ready to screen
- Single tissue rat blots