Multiple tissue blot, single species
Lieferant: G Bioscienes
TB38 TB39
TB38EA 2050 EUR
TB38 TB39
Multiple tissue blot, single species
Elektrophorese-Reagenzien Western Blotting Reagents
These multiple tissue, single species blots permit researchers to visualise the tissue distribution of their protein in a particular species. These have been successfully used for the analysis of a wide variety of proteins. Each blot has multiple tissue types from either human, mouse or rat species.