Brain tissue region blots, single species
Lieferant: G Bioscienes
TB40
TB41
TB57
TB59
TB40EA
2480
EUR
TB40
TB41
TB57
TB59
Brain tissue region blots, single species
Elektrophorese-Reagenzien
Western Blotting Reagents
G-Biosciences offers ready-to-screen, normal tissue, brain tissue region blots. The brain tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse, rat, human, and primate brains.
Premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using Western transfer buffer.