Kidney tissue region blot, single species
Lieferant: G Bioscienes
TB42 TB43
TB42EA 1120 EUR
TB42 TB43
Kidney tissue region blot, single species
Elektrophorese-Reagenzien Western Blotting Reagents
These are ready to screen, normal tissue, kidney tissue region blots from mouse and rat. The kidney tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse and rat kidney.