Eye tissue region blot, single species
Lieferant: G Bioscienes
TB44
TB45
TB44EA
1120
EUR
TB44
TB45
Eye tissue region blot, single species
Elektrophorese-Reagenzien
Western Blotting Reagents
These are ready to screen, normal tissue, eye tissue region blots from rat and rabbit. The eye tissue specific regions are prepared by carefully dissecting out anatomically and functionally distinct regions of adult rat and rabbit eyes.