Optimum growth flask
Lieferant: THOMSON INSTRUMENT
931116
THMS931116EA 667 EUR
THMS931116
Optimum growth flask
Kolben Zellkulturkolben
Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.
  • Baffles designed for high aeration and low shear
  • Same footprint as compairable Fernbach flask
  • Less foaming than disposable Fernbach flask
  • Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
  • 0,2 μm vented cap
  • Individually packaged and sterilised