RBC Lysis Buffer (10X)
Lieferant: Tonbo Biosciences
TNB-4300-L100
TNB-4300-L100EA
41.6
EUR
TNB-4300-L100
RBC Lysis Buffer (10X)
Puffer
Extraktionspuffer
RBC Lysis Buffer (10X) is a concentrated ammonium chloride-based lysing reagent.
- Reactivity: Mouse, Human
The diluted 1X working solution will lyse red blood cells in single cell suspensions with minimal effects on leukocytes. RBC Lysis Buffer (10X) does not contain a fixative so the cells remain viable after red blood cell lysis.