Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
0014-24A-BIOT
USBI0014-24A-BIOTEA 884 EUR
USBI0014-24A-BIOT
Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR