Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: US Biological
0014-24A-HRP
USBI0014-24A-HRPEA 884 EUR
USBI0014-24A-HRP
Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500

Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR