Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody
Lieferant: US Biological
0014-24B
USBI0014-24BEA 667 EUR
USBI0014-24B
Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human