Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
0014-24B-PE
USBI0014-24B-PEEA 884 EUR
USBI0014-24B-PE
Anti-14-3-3 delta/zeta Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!
Type: Primary
Antigen: 14-3-3 delta/zeta (14-3-3 delta/zeta)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human