Anti-5-Methylcytosine Mouse Monoclonal Antibody [clone: 13A149]
Lieferant: US Biological
029406
USBI029406EA 701 EUR
USBI029406
Anti-5-Methylcytosine Mouse Monoclonal Antibody [clone: 13A149]
Antikörper
Recommended dilution: Dot Blot: 1 ?g/ml
Type: Primary
Antigen: 5-Methylcytosine
Clonality: Monoclonal
Clone: 13A149
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human