Anti-Cholesterol Acyltransferase 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
029444
USBI029444EA 769 EUR
USBI029444
Anti-Cholesterol Acyltransferase 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0. 05 ?g/ml; Immunochemistry: 1:1000

Type: Primary
Antigen: Cholesterol Acyltransferase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR