Anti-HIST3H3 Rabbit Polyclonal Antibody

Lieferant: US Biological
029499
USBI029499EA 701 EUR
USBI029499
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR