Anti-TRAF2 Rabbit Polyclonal Antibody

Lieferant: US Biological
029627
USBI029627EA 590 EUR
USBI029627
Anti-TRAF2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.1 ?g/ml; Immunochemistry: 1:500

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR