Anti-PAK7 Rabbit Polyclonal Antibody

Lieferant: US Biological
029860
USBI029860EA 449 EUR
USBI029860
Anti-PAK7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-4?g/ml

Type: Primary
Antigen: PAK7 (p21 protein (Cdc42/Rac)-activated kinase 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR